Abnova
Manufacturer Code:MAB19041
Catalog # 89-335-875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E4 |
Immunogen | A synthetic phosphopeptide corresponding to residues surrounding S380/T382/T383 of human PTEN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 10q23del; BZS; MGC11227; MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; MMAC1; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; Mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin-like protein; Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; PTEN1; TEP1
Gene Aliases: 10q23del; BZS; CWS1; DEC; GLM2; MHAM; MMAC1; PTEN; PTEN1; TEP1
UniProt ID: (Human) P60484
Entrez Gene ID: (Human) 5728
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.