R&D Systems
Manufacturer Code:AF6655SP
Catalog # AF6655SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human PTEN Thr2-Val403 Accession # P60484 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BZS EC 3.1.3.1610q23del EC 3.1.3.48 EC 3.1.3.67 GLM2 MMAC1 MMAC1 phosphatase and tensin homolog deleted on chromosome 10 MMAC1MGC11227 Mutated in multiple advanced cancers 1 phosphatase and tensin homologDEC phosphatidylinositol-345-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN PTEN1 TEP1MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; Mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin-like protein; Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN
Gene Aliases: 10q23del; BZS; CWS1; DEC; GLM2; MHAM; MMAC1; PTEN; PTEN1; TEP1
UniProt ID: (Human) P60484
Entrez Gene ID: (Human) 5728
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.