Novus Biologicals
Manufacturer Code:NBP29300520UL
Catalog # NB027907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 172-281 of human PTCRA (NP_612153.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pre T-cell antigen receptor alpha; pre T-cell antigen receptor alpha PTA PT-ALPHA; pre-T-cell receptor alpha chain; pT-alpha; pT-alpha-TCR
Gene Aliases: PT-ALPHA; PTA; PTCRA
UniProt ID: (Human) Q6ISU1
Entrez Gene ID: (Human) 171558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.