Invitrogen
Manufacturer Code:PA572748
Catalog # PIPA572748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTCHD2 antibody for a 19 amino acid synthetic peptide from near the center of human PTCHD2. The immunogen is within amino acids 780 - 830 of PTCHD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dispatched 3; dispatched homolog 3; patched domain containing 2; Patched domain-containing protein 2; Patched domain-containing protein 2 Protein dispatched homolog 3 PTCHD2 DISP3 KIAA1337; Protein dispatched homolog 3; RND-type protein RNDEu-2
Gene Aliases: DISP3; G630052C06Rik; KIAA1337; PTCHD2; RGD1305820; RNDEu-2
UniProt ID: (Human) Q9P2K9
Entrez Gene ID: (Human) 57540, (Rat) 313705, (Mouse) 242748
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.