Abnova
Manufacturer Code:PAB25704
Catalog # 89-127-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at N-terminus of human PTCHD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PTCHD2.
For Research Use Only
Protein Aliases: DISP3; dispatched 3; dispatched homolog 3; KIAA1337; patched domain containing 2; Patched domain-containing protein 2; Protein dispatched homolog 3
Gene Aliases: DISP3; KIAA1337; PTCHD2
UniProt ID: (Human) Q9P2K9
Entrez Gene ID: (Human) 57540
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.