Invitrogen
Manufacturer Code:PA518544
Catalog # PIPA518544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (HPESRHHPPSNPRQQ) corresponding to the internal amino acids of PTCH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: mesenchymal dysplasia; Patched 1; Protein patched homolog 1; PTC; PTCH patched homolog (Drosophila) RP11-435O5.3 BCNS FLJ42602 HPE7 NBCCS PTC PTC1 PTCH1 PTCH protein +12b PTCH protein +4&prime PTCH protein -10 patched patched (Drosophila) homolog; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3,4,5; Ptch1 protein +12b
Gene Aliases: A230106A15Rik; BCNS; HPE7; mes; NBCCS; PTC; PTC1; PTCH; PTCH1; PTCH11
UniProt ID: (Human) Q13635, (Mouse) Q61115
Entrez Gene ID: (Human) 5727, (Mouse) 19206
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.