Invitrogen
Manufacturer Code:PA123381
Catalog # PIPA123381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues K(269) K I N Y Q V D S W E(279) of mouse PTCH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mesenchymal dysplasia; Patched 1; Patched PTC1 PTC Protein patched homolog 1; Protein patched homolog 1; PTC; Ptch1 protein +12b
Gene Aliases: A230106A15Rik; mes; Ptc; Ptc1; Ptch; Ptch1
UniProt ID: (Mouse) Q61115
Entrez Gene ID: (Mouse) 19206
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.