Invitrogen
Manufacturer Code:PA578547
Catalog # PIPA578547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PTBP2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; neural polypyrimidine tract binding protein; Neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; polypyrimidine tract binding protein 2 PTBLP PTB nPTB8 nPTB6 nPTB nPTB5 nPTB7 brPTB; Polypyrimidine tract-binding protein 2; PTB-like protein; RRM-type RNA-binding protein brPTB
Gene Aliases: Brptb; NPTB; PTB; Ptb2; PTBLP; PTBP2
UniProt ID: (Human) Q9UKA9, (Mouse) Q91Z31, (Rat) Q66H20
Entrez Gene ID: (Human) 58155, (Mouse) 56195, (Rat) 310820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.