Novus Biologicals
Manufacturer Code:NBP219997
Catalog # NBP219997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human PTBP2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brPTB FLJ34897 neural polypyrimidine tract binding protein Neural polypyrimidine tract-binding protein Neurally-enriched homolog of PTB NPTB nPTB6 nPTB7 nPTB8 polypyrimidine tract binding protein 2 polypyrimidine tract-binding protein 2 PTB-like protein PTBPTBLPnPTB5 splicing regulator; neural polypyrimidine tract binding protein; Neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; Polypyrimidine tract-binding protein 2; PTB-like protein
Gene Aliases: brPTB; NPTB; PTB; PTBLP; PTBP2
UniProt ID: (Human) Q9UKA9
Entrez Gene ID: (Human) 58155
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.