Novus Biologicals
Manufacturer Code:NBP30295620UL
Catalog # NB092409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PTBP1 (NP_114368.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 57 kDa RNA-binding protein PPTB-1; Heterogeneous nuclear ribonucleoprotein I; Heterogeneous nuclear ribonucleoprotein I heterogeneous nuclear ribonucleoprotein polypeptide I hnRNP I HNRNPI HNRNP-I HNRPI MGC10830 MGC8461 polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoproteinI) polypyrimidine tract binding protein 1 polypyrimidine tract-binding protein 1 pPTB PTB-1 PTB2 PTB3 PTB4 PTB57 kDa RNA-binding protein PPTB-1 PTB-T RNA-binding protein; heterogeneous nuclear ribonucleoprotein polypeptide I; hnRNP I; polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoprotein I); Polypyrimidine tract-binding protein 1; PTB; RNA-binding protein
Gene Aliases: HNRNP-I; HNRNPI; HNRPI; pPTB; PTB; PTB-1; PTB-T; PTB2; PTB3; PTB4; PTBP1
UniProt ID: (Human) P26599
Entrez Gene ID: (Human) 5725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.