Invitrogen
Manufacturer Code:MA527857
Catalog # PIMA527857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT982 |
Immunogen | Recombit protein encompassing a sequence within the center region of human PSRC1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: differential display and activated by p53; p53-regulated DDA3; proline/serine-rich coiled-coil 1; Proline/serine-rich coiled-coil protein 1
Gene Aliases: DDA3; FP3214; PSRC1
UniProt ID: (Human) Q6PGN9
Entrez Gene ID: (Human) 84722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.