Invitrogen
Manufacturer Code:MA529510
Catalog # PIMA529510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombit Human PSPH protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; Phosphoserine phosphatase; PSP; PSPase; PSPH phosphoserine phosphatase PSP L-3-phosphoserine phosphatase O-phosphoserine phosphohydrolase OTTHUMP00000025059 PSPase
Gene Aliases: PSP; PSPH; PSPHD
UniProt ID: (Human) P78330
Entrez Gene ID: (Human) 5723
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.