Invitrogen
Manufacturer Code:PA519113
Catalog # PIPA519113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RQQVKDNAKWYITD) corresponding to the C-terminus amino acids of PSPH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; Phosphoserine phosphatase; PSP; PSPase; PSPH phosphoserine phosphatase PSP L-3-phosphoserine phosphatase O-phosphoserine phosphohydrolase OTTHUMP00000025059 PSPase
Gene Aliases: AI480570; PSP; PSPase; PSPH; PSPHD
UniProt ID: (Human) P78330, (Mouse) Q5XHW3
Entrez Gene ID: (Human) 5723, (Mouse) 100678
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.