Proteintech
Manufacturer Code:145131AP150UL
Catalog # 50-172-9503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PSPH Fusion Protein Ag5972 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L 3 phosphoserine phosphatase phosphoserine phosphatase PSP PSPase PSPH; L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; Phosphoserine phosphatase; PSP; PSPase
Gene Aliases: AI480570; PSP; PSPase; PSPH; PSPHD
UniProt ID: (Human) P78330, (Mouse) Q5XHW3, (Rat) Q5M819
Entrez Gene ID: (Human) 5723, (Mouse) 100678, (Rat) 304429
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.