Invitrogen
Manufacturer Code:PA518133
Catalog # PIPA518133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DHLPPPGYDDMYL) corresponding to the C-terminus amino acids of PSMF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hPI31; proteasome (prosome, macropain) inhibitor subunit 1 (PI31); Proteasome inhibitor PI31 subunit; PSMF1 PI31 proteasome (prosome macropain) inhibitor subunit 1 (PI31) RP4-545L17.1 proteasome inhibitor hP131 subunit proteasome inhibitor subunit 1
Gene Aliases: PI31; PSMF1
UniProt ID: (Human) Q92530
Entrez Gene ID: (Human) 9491
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.