Proteintech
Manufacturer Code:129411AP150UL
Catalog # 50-172-8363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PSMF1 Fusion Protein Ag4011 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPI31; hPI31 PI31 PSMF1; proteasome (prosome, macropain) inhibitor subunit 1 (PI31); Proteasome inhibitor PI31 subunit; proteasome inhibitor subunit 1
Gene Aliases: AW048666; BC012260; PI31; PSMF1
UniProt ID: (Human) Q92530, (Mouse) Q8BHL8, (Rat) Q5XIU5
Entrez Gene ID: (Human) 9491, (Mouse) 228769, (Rat) 689852
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.