Novus Biologicals
Manufacturer Code:NBP222236H
Catalog # NBP222236H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1 and 50 of human Proteasome (Prosome Macropain) Activator subunit 4 using the numbering given in entry NP_055429.2 (GeneID 23198). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: proteasome (prosome macropain) activator subunit 4; proteasome (prosome, macropain) activator subunit 4; proteasome activator 200 kDa; Proteasome activator complex subunit 4; Proteasome activator PA200
Gene Aliases: KIAA0077; PA200; PSME4
UniProt ID: (Human) Q14997
Entrez Gene ID: (Human) 23198
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.