Invitrogen
Manufacturer Code:PA521789
Catalog # PIPA521789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment contain a sequence corresponding to a region within amino acids 1 and 254 of PSME3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11S regulator complex gamma subunit; 11S regulator complex subunit gamma; Activator of multicatalytic protease subunit 3; epididymis secretory protein Li 283; Ki antigen; Ki nuclear autoantigen; Ki PA28-gamma PA28G REG-GAMMA 11S regulator complex gamma subunit 11S regulator complex subunit gamma Ki antigen Ki nuclear autoantigen PA28gamma activator of multicatalytic protease subunit 3 proteasome activator 28 subunit gamma proteasome activator 28-gamma proteasome activator complex subunit 3; PA28 gamma variant 5; PA28g; proteaseome (prosome, macropain) 28 subunit, 3; proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki); Proteasome activator 28 subunit gamma; proteasome activator 28-gamma; Proteasome activator complex subunit 3; REG gamma-3; REG-gamma
Gene Aliases: AA410043; Ab2-371; AU020960; HEL-S-283; Ki; PA28-gamma; PA28G; PA28gamma; PSME3; REG-GAMMA; REGgamma
UniProt ID: (Human) P61289, (Mouse) P61290
Entrez Gene ID: (Human) 10197, (Mouse) 19192, (Rat) 287716
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.