Invitrogen
Manufacturer Code:PA528039
Catalog # PIPA528039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 238 of Human PSME1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11S regulator complex subunit alpha; 29-kD MCP activator subunit; Activator of multicatalytic protease subunit 1; epididymis secretory sperm binding protein Li 129m; IFI5111 PA28A PA28alpha REGalpha 11S regulator complex alpha subunit 11S regulator complex subunit alpha 29-kD MCP activator subunit IGUP I-5111 REG-alpha activator of multicatalytic protease subunit 1 interferon gamma up-regulated I-5111 protein interferon-gamma IEF SSP 5111 interferon-; IGUP I-5111; Interferon gamma up-regulated I-5111 protein; interferon-gamma IEF SSP 5111; interferon-gamma-inducible protein 5111; PA28a; proteasome (prosome, macropain) activator subunit 1 (PA28 alpha); Proteasome activator 28 subunit alpha; Proteasome activator complex subunit 1; REG-alpha
Gene Aliases: HEL-S-129m; IFI5111; PA28A; PA28alpha; PSME1; REGalpha
UniProt ID: (Human) Q06323
Entrez Gene ID: (Human) 5720
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.