Invitrogen
Manufacturer Code:PA517293
Catalog # PIPA517293
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Lys13 of human PA28a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11S regulator complex alpha subunit 11S regulator complex subunit alpha 29-kD MCP activator subunit IGUP I-5111 OTTHUMP00000164702 REG-alpha activator of multicatalytic protease subunit 1 interferon gamma up-regulated I-5111 protein interferon-gamma IEF SSP 5111 interferon-gamma-inducible protein 5111 proteasome activator 28 subunit alpha proteasome activator complex subunit 1 proteasome activator subunit-1; 11S regulator complex subunit alpha; 29-kD MCP activator subunit; Activator of multicatalytic protease subunit 1; epididymis secretory sperm binding protein Li 129m; IGUP I-5111; Interferon gamma up-regulated I-5111 protein; interferon-gamma IEF SSP 5111; interferon-gamma-inducible protein 5111; PA28 alpha subunit; PA28a; PA28alpha; protease (prosome, macropain) 28 subunit, alpha; proteasome (prosome, macropain) 28 subunit, alpha; proteasome (prosome, macropain) activator subunit 1 (PA28 alpha); Proteasome activator 28 subunit alpha; Proteasome activator complex subunit 1; REG-alpha
Gene Aliases: AW413925; HEL-S-129m; I79_008549; IFI5111; PA28A; PA28alpha; PSME1; REGalpha
UniProt ID: (Human) Q06323, (Mouse) P97371
Entrez Gene ID: (Human) 5720, (Mouse) 19186, (Chinese hamster) 100761736, (Rat) 29630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.