Invitrogen
Manufacturer Code:MA527265
Catalog # PIMA527265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5H2 |
Immunogen | Full length human recombit protein of PSMD9 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome non-ATPase regulatory subunit 9 26S proteasome regulatory subunit p27 Homolog of rat Bridge 1 p27 Proteasome (prosome macropain) 26S subunit non ATPase 9 Proteasome 26S non ATPase regulatory subunit 9 Proteasome 26S subunit non ATPase 9 Psmd9 PSMD9&lowbarHUMAN Rpn4; 26S proteasome regulatory subunit p27; homolog of rat Bridge 1; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9
Gene Aliases: p27; PSMD9; Rpn4
UniProt ID: (Human) O00233
Entrez Gene ID: (Human) 5715
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.