Invitrogen
Manufacturer Code:PA530135
Catalog # PIPA530135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 285 of Human PSMD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit RPN10; 26S proteasome regulatory subunit S5A; AF; AF AF-1 ASF MCB1 Rpn10 S5A pUB-R5 26S proteasome non-ATPase regulatory subunit 4 26S proteasome regulatory subunit S5A RPN10 homolog S5a/antisecretory factor protein angiocidin antisecretory factor 1 multiubiquitin chain-binding protein; angiocidin; Antisecretory factor 1; Multiubiquitin chain-binding protein; proteasome (prosome, macropain) 26S subunit, non-ATPase, 4; S5a/antisecretory factor protein
Gene Aliases: AF; AF-1; ASF; MCB1; PSMD4; pUB-R5; Rpn10; S5A
UniProt ID: (Human) P55036
Entrez Gene ID: (Human) 5710
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.