Invitrogen
Manufacturer Code:PA517259
Catalog # PIPA517259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Gly344 of human S5a/PSMD4 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 26S protease subunit S5a 26S proteasome non-ATPase regulatory subunit 4 26S proteasome regulatory subunit RPN10 26S proteasome regulatory subunit S5A RPN10 homolog S5a/antisecretory factor protein angiocidin antisecretory factor 1 multiubiquitin chain binding protein multiubiquitin chain-binding protein; 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit RPN10; 26S proteasome regulatory subunit S5A; AF; angiocidin; anti secretory factor 1; antisecretory factor; Antisecretory factor 1; Multiubiquitin chain-binding protein; multiubiquitin-chain-binding protein; proteasome (prosome, macropain) 26S subunit, non-ATPase, 4; RPN10 homolog; S5a/antisecretory factor protein
Gene Aliases: AF; AF-1; Af1; angiocidin; ASF; MCB1; PSMD4; pUB-R5; Rpn10; S5A
UniProt ID: (Human) P55036, (Mouse) O35226
Entrez Gene ID: (Human) 5710, (Mouse) 19185, (Rat) 83499
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.