Invitrogen
Manufacturer Code:PA551776
Catalog # PIPA551776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PSMD10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 10; 26S proteasome regulatory subunit p28; ankyrin repeat protein; dJ889N15.2 p28 p28(GANK) 26S proteasome non-ATPase regulatory subunit 10 26S proteasome regulatory subunit p28 ankyrin repeat protein gankyrin hepatocellular carcinoma-associated protein p28-II; Gankyrin; hepatocellular carcinoma-associated protein p28-II; p28(GANK); proteasome (prosome, macropain) 26S subunit, non-ATPase, 10; proteasome subunit p28; proteosome (prosome, macropain) 26S subunit, non-ATPase, 10; proteosome subunit p28
Gene Aliases: AW554874; dJ889N15.2; p28; p28(GANK); p28Gank; PSMD10
UniProt ID: (Human) O75832, (Rat) Q9Z2X3, (Mouse) Q9Z2X2
Entrez Gene ID: (Human) 5716, (Rat) 116722, (Mouse) 53380
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.