Invitrogen
Manufacturer Code:PA543070
Catalog # PIPA543070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PSMC3IP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBD-interacting; Homologous-pairing protein 2 homolog; homologous-pairing protein 2 homolog-like; Nuclear receptor coactivator GT198; proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein; Proteasome 26S ATPase subunit 3-interacting protein; PSMC3 interacting protein GT198 HOP2 TBPIP ODG3 HUMGT198A; PSMC3-interacting protein; Tat-binding protein 1-interacting protein; TBP-1-interacting protein
Gene Aliases: C79099; GT198; HOP2; HUMGT198A; ODG3; PSMC3IP; TBPIP
UniProt ID: (Human) Q9P2W1, (Mouse) O35047, (Rat) Q91ZY6
Entrez Gene ID: (Dog) 607659, (Rabbit) 100344940, (Goat) 100861043, (Bovine) 508530, (Horse) 100052642, (Human) 29893, (Mouse) 19183, (Rat) 140938, (Guinea pig) 100719118
Molecular Function: DNA binding protein enzyme modulator nucleic acid binding signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.