Abnova
Manufacturer Code:H00029893K
Catalog # 89-202-294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PSMC3IP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PSMC3IP peptide using ARM Technology.
For Research Use Only
Protein Aliases: DBD-interacting; GT198 HOP2 HUMGT198A TBPIP; Homologous-pairing protein 2 homolog; Nuclear receptor coactivator GT198; Proteasome 26S ATPase subunit 3-interacting protein; PSMC3-interacting protein; Tat-binding protein 1-interacting protein; TBP-1-interacting protein
Gene Aliases: GT198; HOP2; HUMGT198A; ODG3; PSMC3IP; TBPIP
UniProt ID: (Human) Q9P2W1
Entrez Gene ID: (Human) 29893
Molecular Function: DNA binding protein enzyme modulator nucleic acid binding signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.