OriGene
Manufacturer Code:TA504349
Catalog # 50-167-5065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 21-219 of human PSMB9 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: large multifunctional peptidase 2; LMP2 PSMB6i RING12 beta1i; Low molecular mass protein 2; Macropain chain 7; Multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome catalytic subunit 1i; Proteasome chain 7; proteasome subunit beta 6i; Proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; Really interesting new gene 12 protein
Gene Aliases: beta1i; LMP2; PSMB6i; PSMB9; RING12
UniProt ID: (Human) P28065
Entrez Gene ID: (Human) 5698
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.