Invitrogen
Manufacturer Code:MA535805
Catalog # PIMA535805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1629 |
Immunogen | A synthesized peptide derived from human PSB9 / LMP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: beta1i LMP2 Lmp-2 LMP-2d Low molecular mass protein 2 Macropain chain 7 MGC70470 Multicatalytic endopeptidase complex chain 7 Proteasome chain 7 Proteasome subunit beta type-9 Proteasome subunit beta-1i PSMB6i PSMB9 Really interesting new gene 12 protein Ring12; large multifunctional peptidase 2; large multifunctional protease 2; LMP-2d; low molecular mass polypeptide 2; Low molecular mass protein 2; Macropain chain 7; Multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome catalytic subunit 1i; Proteasome chain 7; proteasome subunit beta 6i; proteasome subunit beta type 9; Proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; proteosome (prosome, macropain) subunit, beta type 9; proteosome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteosome beta 9 subunit; Really interesting new gene 12 protein; RING12 protein
Gene Aliases: beta1i; Lmp-2; LMP2; PSMB6i; PSMB9; RING12
UniProt ID: (Human) P28065, (Rat) P28077, (Mouse) P28076
Entrez Gene ID: (Human) 5698, (Rat) 24967, (Mouse) 16912
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.