Invitrogen
Manufacturer Code:PA519384
Catalog # PIPA519384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RNISYKYRED) corresponding to the internal amino acids of PSMB9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: large multifunctional peptidase 2; large multifunctional protease 2; low molecular mass polypeptide 2; Low molecular mass protein 2; Macropain chain 7; Multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome catalytic subunit 1i; Proteasome chain 7; proteasome subunit beta 6i; Proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; proteosome (prosome, macropain) subunit, beta type 9; PSMB9 proteasome (prosome macropain) subunit beta type 9 (large multifunctional peptidase 2) LMP2 MGC70470 PSMB6i RING12 beta1i low molecular mass protein 2 macropain chain 7 multicatalytic endopeptidase complex chain 7 proteasome beta 9 subunit proteasome catalytic subunit 1i proteasome chain 7 proteasome subunit beta 6i proteasome-related gene 2; Really interesting new gene 12 protein; RING12 protein
Gene Aliases: beta1i; LMP2; PSMB6i; PSMB9; RING12
UniProt ID: (Human) P28065, (Rat) P28077
Entrez Gene ID: (Human) 5698, (Rat) 24967
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.