Invitrogen
Manufacturer Code:PA5102828
Catalog # PIPA5102828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PSMB4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 26 kDa prosomal protein; 26 kDa prosomal protein HN3 HsBPROS26 HsN3 Lmp3 Low molecular mass protein 3 Macropain beta chain Multicatalytic endopeptidase complex beta chain PROS26 PROS-26 Pros-27 proteasome (prosome macropain) subunit beta type 4 proteasome (prosome macropain) subunit beta type 4 proteasome beta 4 subunit proteasome beta chain Proteasome chain 3 proteasome subunit beta 4 proteasome subunit beta type-4 proteasome subunit HsN3 proteasome subunit beta type 4 PSMB4 RN3 testis tissue sperm-binding protein Li 79P; HsBPROS26; HsN3; Low molecular mass protein 3; Macropain beta chain; Multicatalytic endopeptidase complex beta chain; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; Proteasome beta chain; Proteasome chain 3; Proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; RN3; testis tissue sperm-binding protein Li 79P
Gene Aliases: HN3; HsN3; Lmp3; PROS-26; Pros-27; PROS26; PSMB4
UniProt ID: (Human) P28070, (Mouse) P99026
Entrez Gene ID: (Human) 5692, (Rat) 58854, (Mouse) 19172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.