Proteintech
Manufacturer Code:159831AP150UL
Catalog # 50-173-0620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PSMB3 Fusion Protein Ag8784 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20S proteasome HC10 II Proteasome chain 13 Proteasome component C10 II Proteasome subunit beta type 3 Proteasome theta chain PSMB3; proteasome (prosome, macropain) subunit, beta type, 3; proteasome beta 3 subunit; Proteasome chain 13; Proteasome component C10-II; Proteasome subunit beta type-3; Proteasome theta chain
Gene Aliases: AL033320; C10-II; HC10-II; PSMB3
UniProt ID: (Human) P49720, (Mouse) Q9R1P1, (Rat) P40112
Entrez Gene ID: (Human) 5691, (Mouse) 26446, (Rat) 29676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.