Invitrogen
Manufacturer Code:PA556404
Catalog # PIPA556404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PSMB10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Low molecular mass protein 10; Macropain subunit MECl-1; Multicatalytic endopeptidase complex subunit MECl-1; proteasome (prosome, macropain) subunit, beta type, 10; proteasome catalytic subunit 2i; Proteasome MECl-1; proteasome subunit beta 7i; Proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit MECL1; PSMB10 proteasome (prosome macropain) subunit beta type 10 LMP10 MECL1 MGC1665 beta2i macropain subunit MECl-1 multicatalytic endopeptidase complex subunit MECl-1 proteasome MECl-1 proteasome beta 10 subunit proteasome catalytic subunit 2i proteasome subunit MECL1 proteasome subunit beta 7i
Gene Aliases: beta2i; LMP10; MECL1; PSMB10
UniProt ID: (Human) P40306
Entrez Gene ID: (Human) 5699
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.