OriGene
Manufacturer Code:CF800109
Catalog # 50-166-4868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G10 |
Immunogen | Full length human recombit protein of human PSMA6 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa prosomal protein; IOTA PROS27 p27K; Macropain iota chain; macropain subunit iota; Multicatalytic endopeptidase complex iota chain; PROS-27; prosomal P27K protein; proteasome (prosome, macropain) subunit, alpha type, 6; Proteasome iota chain; Proteasome subunit alpha type-6; proteasome subunit iota; testicular secretory protein Li 44
Gene Aliases: IOTA; p27K; PROS27; PSMA6
UniProt ID: (Human) P60900
Entrez Gene ID: (Human) 5687
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.