Proteintech
Manufacturer Code:131631AP150UL
Catalog # 50-172-8535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PSMA/GCPII Fusion Protein Ag3812 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 27 protein; FGCP; FGCP Folate hydrolase 1 FOLH FOLH1 GCP2 GCPII Glutamate carboxypeptidase 2 Glutamate carboxypeptidase II mGCP NAALAD1 NAALAdase NAALADase I PSM PSMA; Folate hydrolase 1; Folylpoly-gamma-glutamate carboxypeptidase; GCPII; glutamate carboxylase II; Glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; Membrane glutamate carboxypeptidase; mGCP; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; NAALADase I; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; Pteroylpoly-gamma-glutamate carboxypeptidase
Gene Aliases: FGCP; FOLH; FOLH1; GCP2; GCPII; GIG27; mGCP; Mopsm; Naalad; NAALAD1; NAALAdase; PSM; PSMA
UniProt ID: (Human) Q04609, (Mouse) O35409, (Rat) P70627
Entrez Gene ID: (Human) 2346, (Mouse) 53320, (Rat) 85309
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.