Novus Biologicals
Manufacturer Code:NBP145058
Catalog # NBP145058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GCP-05 |
Immunogen | amino acids 44-750 of human GCPII |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 27 protein; Cell growth-inhibiting gene 27 protein cell growth-inhibiting protein 27 EC 3.4.17.21 folate hydrolase (prostate-specific membrane antigen) 1 Folate hydrolase 1 FOLHNAALADase I Folylpoly-gamma-glutamate carboxypeptidase GCP2 GCPIImGCP glutamate carboxylase II glutamate carboxypeptidase 2 Glutamate carboxypeptidase II Membrane glutamate carboxypeptidase NAALAD1N-acetylated-alpha-linked acidic dipeptidase I NAALAdase N-acetylated alpha-linked acidic dipeptidase 1 prostate specific membrane antigen variant F Prostate-specific membrane antigen PSMAFGCP PSMPteroylpoly-gamma-glutamate carboxypeptidase; FGCP; Folate hydrolase 1; Folylpoly-gamma-glutamate carboxypeptidase; GCPII; glutamate carboxylase II; Glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; Membrane glutamate carboxypeptidase; mGCP; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; NAALADase I; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; PSM; Pteroylpoly-gamma-glutamate carboxypeptidase
Gene Aliases: FGCP; FOLH; FOLH1; GCP2; GCPII; GIG27; mGCP; NAALAD1; NAALAdase; PSM; PSMA
UniProt ID: (Human) Q04609
Entrez Gene ID: (Human) 2346
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.