R&D Systems
Manufacturer Code:FAB4234A
Catalog # FAB4234A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 460420 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human PSMA/FOLH1/NAALADase I Lys44-Ala750 Accession # Q04609 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 27 protein; Cell growth-inhibiting gene 27 protein cell growth-inhibiting protein 27 EC 3.4.17.21 FGCP folate hydrolase (prostate-specific membrane antigen) 1 Folate hydrolase 1 FOLH1 FOLHNAALADase I Folylpoly-gamma-glutamate carboxypeptidase GCP2 GCPII GCPIImGCP glutamate carboxylase II glutamate carboxypeptidase 2 Glutamate carboxypeptidase II Membrane glutamate carboxypeptidase mopsm NAALAD1 NAALAD1N-acetylated-alpha-linked acidic dipeptidase I NAALAdase NAALADase I N-acetylated alpha-linked acidic dipeptidase 1 prostate specific membrane antigen variant F Prostate-specific membrane antigen PSMAFGCP PSMPteroylpoly-gamma-glutamate carboxypeptidase; FGCP; Folate hydrolase 1; Folylpoly-gamma-glutamate carboxypeptidase; GCPII; glutamate carboxylase II; Glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; Membrane glutamate carboxypeptidase; mGCP; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; NAALADase I; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; PSM; Pteroylpoly-gamma-glutamate carboxypeptidase
Gene Aliases: FGCP; FOLH; FOLH1; GCP2; GCPII; GIG27; mGCP; NAALAD1; NAALAdase; PSM; PSMA
UniProt ID: (Human) Q04609
Entrez Gene ID: (Human) 2346
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.