Invitrogen
Manufacturer Code:MA514821
Catalog # PIMA514821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E.493.5 |
Immunogen | Synthetic peptide corresponding to amino acids surrounding Lys461 of human LEDGF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CLL-associated antigen KW-7; dense fine speckles 70; Dense fine speckles 70 kDa protein; DFS 70; Lens epithelium-derived growth factor; mLEDGF; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52
Gene Aliases: AA408851; AU015605; DFS70; LEDGF; Ledgfa; Ledgfb; p52; p75; PAIP; PSIP1; PSIP2; SBP75
UniProt ID: (Human) O75475, (Mouse) Q99JF8, (Rat) Q812D1
Entrez Gene ID: (Human) 11168, (Mouse) 101739, (Rat) 313323
Molecular Function:
growth factor
signaling molecule
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.