Invitrogen
Manufacturer Code:PA5107009
Catalog # PIPA5107009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: AADRKRKQEE QMETEQQNKD EGKKPEVKKV EKKRETSMDS RLQRIHAEIK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLL-associated antigen KW-7; dense fine speckles 70; Dense fine speckles 70 kDa protein; DFS 70; Lens epithelium-derived growth factor; mLEDGF; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52
Gene Aliases: AA408851; AU015605; DFS70; LEDGF; Ledgfa; Ledgfb; p52; p75; PAIP; PSIP1; PSIP2
UniProt ID: (Human) O75475, (Mouse) Q99JF8
Entrez Gene ID: (Human) 11168, (Mouse) 101739
Molecular Function:
growth factor
signaling molecule
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.