Invitrogen
Manufacturer Code:PA575357
Catalog # PIPA575357
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 202-250 of Human PSGR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled receptor RA1c; GA_x6K02T2PBJ9-5459657-5458695; HPRAJ; Olfactory receptor 51E2; Olfactory receptor 51E2 HPRAJ Olfactory receptor OR11-16 Prostate-specific G-protein coupled receptor OR51E2 PSGR; Olfactory receptor 59; Olfactory receptor 78; olfactory receptor MOR18-2; Olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; prostate specific G-protein coupled receptor; Prostate-specific G-protein coupled receptor
Gene Aliases: 4633402A21Rik; MOL2.3; MOR18-2; Olfr78; Olr59; OR51E2; OR51E3P; OR52A2; PSGR; RA1c
UniProt ID: (Human) Q9H255, (Mouse) Q8VBV9, (Rat) O88628
Entrez Gene ID: (Human) 81285, (Mouse) 170639, (Rat) 170816
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.