Invitrogen
Manufacturer Code:PA534055
Catalog # PIPA534055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from N-terminal extracellular domain of human OR51E2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPRAJ; Olfactory receptor 51E2; Olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; OR51E2 OR51E3P OR52A2 HPRAJ Olfactory receptor 51E2 Olfactory receptor OR11-16 PSGR; prostate specific G-protein coupled receptor; Prostate-specific G-protein coupled receptor
Gene Aliases: OR51E2; OR51E3P; OR52A2; PSGR
UniProt ID: (Human) Q9H255
Entrez Gene ID: (Human) 81285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.