Invitrogen
Manufacturer Code:PA5104387
Catalog # PIPA5104387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human OR51E2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4633402A21Rik GA_x6K02T2PBJ9-5459657-5458695 G-protein coupled receptor RA1c hCG_2044604 HPRAJ MOL2.3 MOR18-2 Olfactory receptor 51E2 olfactory receptor 59 Olfactory receptor 78 olfactory receptor family 51 subfamily E member 2 olfactory receptor MOR18-2 olfactory receptor OR11-16 olfactory receptor family 51 subfamily E member 2 olfactory receptor family 51 subfamily E member 3 pseudogene olfactory receptor family 52 subfamily A member 2 Olfr78 Olr59 OR51E2 OR51E3P OR52A2 prostate specific G-protein coupled receptor prostate-specific G-protein coupled receptor PSGR RA1c; G-protein coupled receptor RA1c; HPRAJ; Olfactory receptor 51E2; Olfactory receptor 59; olfactory receptor 78; Olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; prostate specific G-protein coupled receptor; Prostate-specific G-protein coupled receptor
Gene Aliases: Olfr78; Olr59; OR51E2; OR51E3P; OR52A2; PSGR; RA1c
UniProt ID: (Human) Q9H255, (Rat) O88628
Entrez Gene ID: (Human) 81285, (Rat) 170816
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.