Novus Biologicals
Manufacturer Code:NBP286757
Catalog # NB034379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human PSGR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPRAJ; HPRAJ Olfactory receptor OR11-16 olfactory receptor family 51 subfamily E member 2 olfactory receptor family 51 subfamily E member 3 pseudogene olfactory receptor family 52 subfamily A member 2 OR51E3P OR52A2 prostate specific G-protein coupled receptor Prostate-specific G-protein coupled receptor PSGRolfactory receptor 51E2; Olfactory receptor 51E2; Olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; prostate specific G-protein coupled receptor; Prostate-specific G-protein coupled receptor
Gene Aliases: OR51E2; OR51E3P; OR52A2; PSGR
UniProt ID: (Human) Q9H255
Entrez Gene ID: (Human) 81285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.