Proteintech
Manufacturer Code:161681AP150UL
Catalog # 50-173-0767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AD3L AD3LP AD4 AD5 E5 1 Presenilin 2 PS 2 PS2 PSEN2 PSNL2 STM 2 STM2; AD3LP; AD5; Alzheimer disease 4; E5-1; Presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS-2; STM-2
Gene Aliases: AD3L; AD4; Ad4h; ALG-3; Alg3; CMD1V; PS-2; PS2; PSEN2; PSNL2; STM2
UniProt ID: (Human) P49810, (Mouse) Q61144
Entrez Gene ID: (Human) 5664, (Mouse) 19165
Molecular Function:
aspartic protease
calcium-binding protein
hydrolase
membrane-bound signaling molecule
protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.