Invitrogen
Manufacturer Code:PA585780
Catalog # PIPA585780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse PSD93. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A330103J02Rik antibody B230218P12Rik antibody B330007M19Rik antibody Dlg2 antibody Dlgh2 antibody Gm1197 antibody PSD93 antibody PSD93 antibody discs large homolog 2 (Drosophila) antibody; Channel-associated protein of synapse-110; Chapsyn-110; discs large homolog 2; Disks large homolog 2; Postsynaptic density protein PSD-93; synaptic density protein PSD-93
Gene Aliases: A330103J02Rik; B230218P12Rik; B330007M19Rik; Dlg2; Dlgh2; Gm1197; PSD93
UniProt ID: (Mouse) Q91XM9, (Rat) Q63622
Entrez Gene ID: (Mouse) 23859, (Rat) 64053
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.