Novus Biologicals
Manufacturer Code:NB300546
Catalog # NB300546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues N(352) K L C D K P A S P R H Y S P(366) of human PSD-93. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Channel-associated protein of synapse-110; channel-associated protein of synapses, 110kDa; Chapsyn-110; Chapsyn-110 discs large homolog 2 (Drosophila) discs large homolog 2 chapsyn-110 discs large homolog 2 chapsyn-110 (Drosophila) disks large homolog 2 DKFZp781D1854 DKFZp781E0954 FLJ37266 MGC131811 Postsynaptic density protein PSD-93 PSD-93 PSD93110kDa; discs large 2; discs large homolog 2; discs, large homolog 2; discs, large homolog 2, chapsyn-110; Disks large homolog 2; Postsynaptic density protein PSD-93; protein phosphatase 1, regulatory subunit 58
Gene Aliases: chapsyn-110; DLG2; PPP1R58; PSD-93; PSD93
UniProt ID: (Human) Q15700
Entrez Gene ID: (Human) 1740
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.