Invitrogen
Manufacturer Code:MA531580
Catalog # PIMA531580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1436 |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse PSD95. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 4; Disks large homolog 4; Dlg4 antibody Dlgh4 antibody PSD95 antibody SAP90 antibody SAP90A antibody discs large homolog 4 (Drosophila) antibody; Postsynaptic density protein 95; PSD-95; PSD-95 alpha 2b; PSD-95 beta; SAP-90; Synapse-associated protein 90; synapse-associated protein SAP90
Gene Aliases: Dlg4; Dlgh4; PSD-95; PSD95; SAP90; SAP90A
UniProt ID: (Rat) P31016, (Mouse) Q62108
Entrez Gene ID: (Rat) 29495, (Mouse) 13385
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.