Novus Biologicals
Manufacturer Code:NBP197531
Catalog # NBP197531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7E3-1B8 |
Immunogen | Recombinant rat PSD-95. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 4; discs large homolog 4 (Drosophila) disks large homolog 4 FLJ97752 FLJ98574 post-synaptic density protein 95 Postsynaptic density protein 95 PSD-95 PSD95SAP90SAP-90discs large homolog 4 Synapse-associated protein 90 Tax interaction protein 15; Disks large homolog 4; post-synaptic density protein 95; Postsynaptic density protein 95; PSD-95; SAP-90; Synapse-associated protein 90; Tax interaction protein 15
Gene Aliases: DLG4; PSD95; SAP-90; SAP90
UniProt ID: (Human) P78352
Entrez Gene ID: (Human) 1742
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.