Invitrogen
Manufacturer Code:PA519114
Catalog # PIPA519114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EEQARKAFDRATK) corresponding to the internal amino acids of DLG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 4; Disks large homolog 4; DLG4 discs large homolog 4 (Drosophila) FLJ97752 FLJ98574 PSD95 SAP90 Tax interaction protein 15 discs large homolog 4 post-synaptic density protein 95 synapse-associated protein 90; Postsynaptic density protein 95; PSD-95; SAP-90; Synapse-associated protein 90; synapse-associated protein SAP90
Gene Aliases: Dlg4; Dlgh4; PSD95; Sap90
UniProt ID: (Rat) P31016
Entrez Gene ID: (Rat) 29495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.