Invitrogen
Manufacturer Code:PA576995
Catalog # PIPA576995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PSAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endometrial progesterone-induced protein; EPIP; Phosphohydroxythreonine aminotransferase; Phosphoserine aminotransferase; Phosphoserine aminotransferase Phosphohydroxythreonine aminotransferase PSAT PSAT1 PSA; PSAT
Gene Aliases: D8Ertd814e; EPIP; NLS2; PSA; Psa1; PSAT; PSAT1; PSATD
UniProt ID: (Human) Q9Y617, (Mouse) Q99K85
Entrez Gene ID: (Human) 29968, (Rat) 293820, (Mouse) 107272
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.