Proteintech
Manufacturer Code:183961AP150UL
Catalog # 50-173-2140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PSAP Fusion Protein Ag13263 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1 activator; A1 activator Cerebroside sulfate activator Co beta glucosidase Component C CSAct Dispersin GLBA Glucosylceramidase activator Proactivator polypeptide prosaposin Protein C PSAP SAP 1 SAP 2 Saposin D SAP1; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; Glucosylceramidase activator; Proactivator polypeptide; Prosaposin; Protein A; Protein C; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; SGP-1; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; Sulfated glycoprotein 1; Sulfatide/GM1 activator
Gene Aliases: AI037048; GLBA; PSAP; SAP1; SGP-1; Sgp1
UniProt ID: (Human) P07602, (Mouse) Q61207
Entrez Gene ID: (Human) 5660, (Mouse) 19156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.